Imprimer

Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Biorbyt
ORB27726-0.5 ORB27726-1
BIRBORB27726-0.5EA 0 EUR
BIRBORB27726-0.5 BIRBORB27726-1
Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Type: Primary
Antigen: Avidin
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR