Imprimer

Anti-PARP1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB13638-100 ORB13638-200
BIRBORB13638-100EA 0 EUR
BIRBORB13638-100 BIRBORB13638-200
Anti-PARP1 Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR