Imprimer
Anti-ANK3 Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
ORB13248-100 ORB13248-200
BIRBORB13248-100EA 0 EUR
BIRBORB13248-100 BIRBORB13248-200
Anti-ANK3 Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: ANK3 (ankyrin 3, node of Ranvier (ankyrin G))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant