Imprimer

Anti-TH Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB11539-100 ORB11539-200
BIRBORB11539-100EA 0 EUR
BIRBORB11539-100 BIRBORB11539-200
Anti-TH Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR