Imprimer

Anti-DOPA decarboxylase Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB10526-100 ORB10526-200
BIRBORB10526-100EA 0 EUR
BIRBORB10526-100 BIRBORB10526-200
Anti-DOPA decarboxylase Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: DOPA decarboxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR