Imprimer

Anti-ASTE1 Rabbit polyclonal antibody

Fournisseur: Biorbyt
ORB2022-100 ORB2022-200
BIRBORB2022-100EA 0 EUR
BIRBORB2022-100 BIRBORB2022-200
Anti-ASTE1 Rabbit polyclonal antibody
Anticorps
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: ASTE1 (asteroid homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR