Imprimer

Anti-TTC33 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB1476-100 ORB1476-200
BIRBORB1476-100EA 0 EUR
BIRBORB1476-100 BIRBORB1476-200
Anti-TTC33 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: TTC33 (tetratricopeptide repeat domain 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR