Imprimer
Beakers, low form, Griffin, PYREX® VISTA™
Fournisseur: Corning
70000-250 70000-600
CORN70000-250EA 0 EUR
CORN70000-250 CORN70000-600
Beakers, low form, Griffin, PYREX® VISTA™
Béchers
These beakers with spout are made of Type I Class A low expansion borosilicate glass, per ASTM E438.
  • High mechanical strength
  • Resistance to thermal shock
Beakers have blue graduations and an extra large blue marking spot. The 250 through 1000 ml beakers have a double graduated metric scale to indicate approximate content.
*10 ml beaker is not graduated.
Certifications: ASTM E-960