Imprimer
Deflected point septum penetration needles
Fournisseur: CADENCE SCIENCE
4181 4850 4194 4170 4187 4192 4195 4196 4188
POPR4181EA 932 EUR
POPR4181 POPR4850 POPR4194 POPR4170 POPR4187 POPR4192 POPR4195 POPR4196 POPR4188
Deflected point septum penetration needles
Aiguilles pour seringues
These deflected needle style features point positioning on the axis of the needle lumen. This improved point geometry provides easier penetration and minimises coring and tracking (particularly with finer gauge needles).
Note: Needles not intended for human use.