Imprimer
Carboxyl reactive biotin reagents, HOOK™
Fournisseur: G Bioscienes
BG-16 BG-17
BG-16EA 943 EUR
BG-16 BG-17
Carboxyl reactive biotin reagents, HOOK™
Systèmes de liaison et de marquage des protéines
HOOK™ Biotin-PEG₂-Amine, and its long chain form, HOOK™ Biotin-PEG₃-Amine, are carboxyl reactive biotinylation reagents. These agents contain terminal amines and react with carboxyl groups found at the carboxyl termini, aspartate, and glutamate side chains. The reaction is mediated by a water-soluble carbodiimide (EDC). The carbodiimide activates the carboxyl group and reacts with the amines (-NH₂) on the biotinylation agent to form an amide bond. This reaction is rapid and takes just a few minutes to complete. Under these conditions, hydrazide-derivatives of biotin reagents may also react with the carboxyls.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44c6a50178dcf6735c804a8b8b2d178dc201f96f&vid=b507ed9580523f3ad45b8e5bbfcd53839cbc95af&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85b66c4bf20e86410c6d229693e1b4ec272a1868&vid=e44b9a1ad71eeeacbf9796d4ce8877556e4fcd16&prid=web&clid=SAPDAM