Imprimer
Examination gloves, latex, Gentle Skin® Sensitive
Fournisseur: MEDITRADE
1221T-M 1221T-XL 1221T-L
ROES1221T-MEA 0 EUR
ROES1221T-M ROES1221T-XL ROES1221T-L
Examination gloves, latex, Gentle Skin® Sensitive
Gants
These ambidextrous gloves are designed for use in areas with high contamination risks. Ideal for people working in hospitals, care facilities and laboratories.
  • Non sterile, powder-free, micro-textured finish
  • High elasticity, flexibility and tactility
  • ProSlide effect for easy and quick donning
  • Beaded cuffs
  • Low in proteins and accelerators
Colour: White
EN 455; AQL 1,5
Certifications: EN 455; AQL 1,5