Imprimer
Scalpel blades, carbon steel, Feather®
Fournisseur: PFM Medical GmbH
201500015 201500011 201500024 201500023 201500022 201500021 201500020 201500010
FEAT201500015EA 27.4 EUR
FEAT201500015 FEAT201500011 FEAT201500024 FEAT201500023 FEAT201500022 FEAT201500021 FEAT201500020 FEAT201500010
Scalpel blades, carbon steel, Feather®
Bistouris et pièces détachées Lames de bistouri
Carbon steel, sterile.
  • With anti-rust coating
  • Suitable for all surgical applications
  • Optimised micrometer-accurate precision grinding
  • Double-honing process ensures ultra-sharp cutting edges of superb quality and durability
  • Blade is ready for use immediately after removal from the package and is easy to insert into the handle