Imprimer
Hand cleansing cream, Kresto® paint paste
Fournisseur: SC Johnson
Kresto®
EVON81907A06EA 301 EUR
EVON81907A06
Hand cleansing cream, Kresto® paint paste
Soin de la peau Savons et soins nettoyants
Kresto® paint paste is a non-soapy, viscous, slightly perfumed suspension special hand cleanser with a very powerful cleansing action for the removal of stubborn contamination such as lacquers, resins, adhesives or oil-based paints.
  • Contains active agent for easy removal of inks and dyes from the skin
  • Astopon® natural scrubbing agent (refined walnut shell powder) gently removes contaminants without damaging the skin
  • As the formulation is concentrated, only a small amount is required to remove contaminants
  • Solvent-free, colourant-free, preservative-free, perfumed
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904