Imprimer
Electronic rotary microtome, HM 340 E
Fournisseur: Epredia
SHAN905190EA 0 EUR
SHAN905190
Electronic rotary microtome, HM 340 E
Microtomes et groupes de microtomes Microtomes
Designed for precision and stability, it yields quality ribbons even for difficult to cut sections. Applications range from paraffin sectioning in routine and research laboratories to plastic embedded specimens. The integrated waste tray captures excess paraffin debris to the rear of the blade holder for quick and easy cleaning. To guarantee safe specimen transfer the hand wheel brake locks in any position and the protective blade guard allows operation without exposing a blade edge.
Information de livraison: Supplied with 100 ml PARA/Gard™, standard tools, brushed aluminium plate cover, dust cover, and instruction manual.