Imprimer
Reversed phase HPLC columns, Atlantis® T3
Fournisseur: WATERS
186003728 186003723 186003726 186003727 186003736 186003747 186003719 186003729
WATE186003728EA 905 EUR
WATE186003728 WATE186003723 WATE186003726 WATE186003727 WATE186003736 WATE186003747 WATE186003719 WATE186003729
Reversed phase HPLC columns, Atlantis® T3
Colonnes pour chromatographie
Atlantis T3 columns are universal, silica based, reversed phase C18 columns that provide balanced retention of polar and hydrophobic molecules and useful for developing separations of polar and non-polar compounds.