Imprimer
Analytical HPLC columns, SymmetryShield™ RP8
Fournisseur: WATERS
WAT094260 WAT094269 WAT200670
WATEWAT094260EA 765 EUR
WATEWAT094260 WATEWAT094269 WATEWAT200670
Analytical HPLC columns, SymmetryShield™ RP8
Colonnes pour chromatographie
SymmetryShield™ columns feature Waters' patented embedded polar group technology that shields the silica's residual silanols from highly basic analytes at improves overall peak shape. Additionally, by placing the embedded polar group close to the silica surface, the activity of the surface silanols is further reduced. This imparts selectivity and retention that is different compared to the Symmetry® C18 ligand.