Imprimer
BioHPLC columns, DNAPac™ PA200
Fournisseur: Thermo Fisher Scientific
063419 063421 063423 063425 088780 088781 062998 063000
555-7283EA 1080.2 EUR
555-7283 555-7284 555-7285 555-7286 555-0253 555-7456 555-7323 555-7324
BioHPLC columns, DNAPac™ PA200
Colonnes pour chromatographie
The PA200 columns are strong anion exchange column developed to provide unsurpassed high resolution analysis and purification of synthetic oligonucleotides.
  • With secondary structures it resolve oligonucleotides
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt and solvent
  • pH range: 2 to 12,5