Imprimer
Rotary valves with cap connection
Fournisseur: OMNIFIT
001113
OMNI001113EA 0 EUR
OMNI001113
Rotary valves with cap connection
Vannes Vannes de commutation
Valves are constructed from PTFE with a PCTFE rotor and have a 1,5 mm bore size. They accept tubing sizes between 0,5 and 4 mm Ext.Ø. and are pressure rated to 50 psi.