Imprimer
Cleaning reagents, COULTER CLENZ®
Fournisseur: Beckman Coulter
8448222 8448188
BECL8448222EA 0 EUR
BECL8448222 BECL8448188
Cleaning reagents, COULTER CLENZ®
Produits nettoyants Détergents et Nettoyants de Surfaces
COULTER CLENZ® is an all-purpose liquid cleaning reagent. It contains agents that strip away protein build-up and lysed red blood cells in instrument fluidics. It should be used routinely as part of regular instrument maintenance, such as shutdown. Additionally, it should be included in a schedule of routine preventive maintenance procedures designed to maintain high performance.