Imprimer
Capillary columns, OPTIMA® 5 MS Accent
Fournisseur: MACHEREY-NAGEL
725820.30 725811.60 725820.60 725810.25 725815.12 725820.15 725825.30 725825.60 725826.30 725826.60 725811.30 725813.30 725812.25 725812.60 725810.50 725815.50
MANA725820.30EA 800 EUR
MANA725820.30 MANA725811.60 MANA725820.60 MANA725810.25 MANA725815.12 MANA725820.15 MANA725825.30 MANA725825.60 MANA725826.30 MANA725826.60 MANA725811.30 MANA725813.30 MANA725812.25 MANA725812.60 MANA725810.50 MANA725815.50
Capillary columns, OPTIMA® 5 MS Accent
Colonnes pour chromatographie
Nonpolar silarylene GC phase with low bleeding.
  • Ideal for ion trap and quadrupol MS detectors
  • Widely used for environmental analyses, trace analyses, EPA methods, pesticides, PCB, food and drug analyses
  • Max. temperature for isothermal operation 340 °C
  • Max. temperature for short isotherms in a temperature program 360 °C
Columns with films >0,5 µm: max. temperatures 320 and 340 °C, respectively.
The polarity is similar to 5% diphenyl - 95% dimethylpolysiloxane phases.
USP code G27 / G36