Imprimer
Laboratory bottles, narrow neck, with screw cap
Fournisseur: SMITH SCIENTIFIC
2070/250 2070/5000 2070/100
SMIT2070/250EA 21.8 EUR
SMIT2070/250 SMIT2070/5000 SMIT2070/100
Laboratory bottles, narrow neck, with screw cap
Flacons Flacons en verre
SIMAX®, Borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With screw cap and pouring ring.