Imprimer
Belt, soft buckle, 9342 STRE
Fournisseur: FRISTADS KANSAS
100909940L 100909940M 100909940S
KWIN100909940LEA 24.9 EUR
KWIN100909940L KWIN100909940M KWIN100909940S
Belt, soft buckle, 9342 STRE
Tenue de travail
This belt made of 100% polyester features a soft buckle.
  • For car industry, spraying works etc.
  • Soft buckle in leather and plastic
  • Velcro fastening
  • Width 40 mm