Imprimer
Sodium dodécyle sulfate (SDS), Pierce™
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
28365 28364
PIER28365EA 329 EUR
PIER28365 PIER28364
Sodium dodécyle sulfate (SDS), Pierce™
Sodium dodécyle sulfate (SDS)
Thermo Scientific Sodium Dodecyl Sulfate (Lauryl) is standard-grade SDS detergent for use in protein polyacrylamide gel electrophoresis (PAGE).
  • Popular anionic detergent for a variety of protein methods
  • Especially useful for denaturing polyacrylamide gel electrophoresis
  • Common component of cell lysis buffers
Properties of SDS (values for pure C12):
Detergent Class: Ionic (anionic)
Aggregation Number: 62
Micelle Molecular Weight: 18,000 g
Critical Micelle Concentration (CMC): 6 to 8 mM (0.1728 to 0.2304%, w/v)
Cloud Point: >100°C
Dialyzable: No
Informations de commande: Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Attention: For Research Use Only. Not for use in diagnostic procedures.
Formule: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Point de fusion: 204…207 °C
Densité: 1.1 g/cm³ (20 °C)
Point d'éclair: >100 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00036175
Numéro CAS: 151-21-3
EINECS: 205-788-1
ONU: 1325
ADR: 4.1 II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM