Imprimer
Sodium dodécyle sulfate (SDS) solution_SC 10%, ultrapure
Fournisseur: Thermo Fisher Scientific
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
J77504.K2 J77504.AE J77504.AP
J77504.K2EA 167 EUR
J77504.K2 J77504.AE J77504.AP
Sodium dodécyle sulfate (SDS) solution_SC 10%, ultrapure
Sodium dodécyle sulfate (SDS)
10% (w/v) solution of sodium dodecyl sulfate (SDS) in high purity dH2O. Solution is 0.2μm filtered and dispensed into durable, square Nalgene bottles.
A(280 nm, 3%, 1 cm): ≤ 0.1
A(230 nm, 3%, 1 cm): ≤ 0.2
Attention: Note: Precipitation of SDS is not unusual. Warm gently to redissolve.
Formule: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Densité: 1…1.1 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00036175
Numéro CAS: 151-21-3
EINECS: 205-788-1
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM