Imprimer
Blood collecting system, S-Monovette® Neutral
Fournisseur: SARSTEDT
02.1726.001 01.1728.001 02.1726.021 04.1926.001 05.1727.001 05.1729.001
SARS02.1726.001EA 98 EUR
SARS02.1726.001 SARS01.1728.001 SARS02.1726.021 SARS04.1926.001 SARS05.1727.001 SARS05.1729.001
Blood collecting system, S-Monovette® Neutral
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
The S-Monovette combines two blood collection techniques – The aspiration technique and the vacuum technique.
Suitable for all vein conditions to achieve an optimal sample quality.