Imprimer
Dishes with lid
Fournisseur: Nickel-Electro
NICK503543EA 72.9 EUR
NICK503543
Dishes with lid
Plateaux/Récipients/Pots Plateaux et récipients de laboratoire
Made of nickel.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean