Imprimer
HPLC columns, NUCLEOSIL® Protect I
Fournisseur: MACHEREY-NAGEL
721157.30 720170.40 720170.46 720174.46 720175.40 720175.46
MANA721157.30EA 153 EUR
MANA721157.30 MANA720170.40 MANA720170.46 MANA720174.46 MANA720175.40 MANA720175.46
HPLC columns, NUCLEOSIL® Protect I
Colonnes pour chromatographie
NUCLEOSIL® Protect I is an endcapped alkyl modified silica phase with an embedded polar group, using for HPLC.
  • Robust spherical silica gel
  • Hydrophobic RP phase with polar properties that is suited for 100% aqueous eluents
  • Separation mechanism based on hydrophobic and polar interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).