Imprimer
Masterflex® Variable Area Flowmeter with High-Res Valve, Correlated Reading, High-Flow, 316 Stainless Steel Housing and Fittings; 65-mm Scale; 42.094 LPM Air
Fournisseur: Avantor Fluid Handling
VWR Item#MFLX32046-39
Your Organization's Price:
Read more easily with 16% magnification of scale.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
More Product Information
Other Products In this Family
See all Items in this Product Family
Learn more
About VWR
Plus About VWR