Imprimer
Sanitary 3A Centrifugal Pump, 105 GPM, 1.5 HP, White Epoxy Motor, 3600 RPM
Fournisseur: Avantor Fluid Handling
VWR Item#MFLX76710-10
Your Organization's Price:
Clean-in-place (CIP) 3A sanitary transfer pump.
  • Washdown motors available in white epoxy
  • Stainless steel adapters are standard
  • Ideal for RO water and pharmaceutical manufacturing
Piping changes are eliminated due to the interchangeable design that these economical pumps offer when compared to other pumps. Models are available with inlets up to 6" contact our Application Specialists for assistance in sizing the proper pump for your system.
<b>Information de livraison: </b>Includes conduit box for hardwiring.
More Product Information
Other Products In this Family
See all Items in this Product Family
Learn more
About VWR
Plus About VWR