Masterflex® L/S® Pump Head Adapter for NEMA 56C Frame Motor, 3.7:1 Gear Ratio
Fournisseur: Avantor Fluid Handling
Customize your pump—mount L/S® pump heads to a 56C frame motor.
- Use up to 2 L/S® pump heads—pump two fluids simultaneously at equal or different flow rates
- IP56-rated cast-aluminum adapters feature built-in gear box
- Flow range 0.06 to 3400 mL/min—flow rate depends on drive rpm, tubing head, and tubing size
These adapters easily mount Masterflex® L/S® pump heads to a 56C frame motor (pump head and motor sold separately). Adapters accept seven different pump head types: Standard, Easy-Load®, Easy-Load ® II, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm<br /><br />Use the following formula to calculate flow rate— do not exceed 600 rpm pump head speed: Flow rate = (Motor rpm/adapter gear ratio) x flow per revolution
<b>Information de livraison: </b>Includes mounting hardware.