Imprimer
Masterflex® Mass Flow Controller Power Supply; 110 VAC to 12 VDC
Fournisseur: Avantor Fluid Handling
VWR Item#MFLX32498-40
Your Organization's Price:
Make the most of your Masterflex flowmeters with these replacement parts and other accessories.
  • Optimize performance of your flowmeter with these accessories
  • Provides an easy connection to the flow sensor to facilitate calibration
  • Software makes it easy to finely tune the flow sensor for the flow range that the application needs
  • Enables data to be recorded / captured for real-time observation using a connected computer or download to review at a later time
These power supplies, adapters, and other accessories are suitable for use with your Masterflex flowmeter and flow sensors.
The USB data converter is used to communicate via the serial interface to the Masterflex Ultrasonic Flow Sensor using a computer. The same data cable can used on any of the Masterflex Flow Sensors.<br /><br />The software / data cable enable simple / fine adjustment of the flow sensors to best match it to a specific task. <br /><br />To assist in problem solving, one can view real-time data in a graphical display when connected to a computer or save data points for later analysis. <br /><br />Allows one to use the sensor on a different tubing formulation or wall thickness if desired.<br /><br />Supported operating systems:<br /> - Win98, Win2000, Win XP, Win7, Win8, Win10<br />Screen resolution:<br /> - 1024 x 640 or higher<br />Interface:<br /> -1 USB port<br />Processor:<br /> - Pentium 700 MHz or higher<br />RAM:<br /> - 256 MB or higher<br />Software (needed by user - not provided):<br /> - Excel (for the management of the measurement data)
<b>Information de livraison: </b>Includes specially adapted cable (has one USB for both data and power and a second USB for additional power - if needed - on one side and on the other is a M12 connector for attaching to the Masterflex Flow Sensor) ; and software on a USB drive.
More Product Information
Other Products In this Family
See all Items in this Product Family
Learn more
About VWR
Plus About VWR