Imprimer
Micropump L21837 Gear Pump Accessory Service Kit for 07003-04; 1/Ea
Fournisseur: Avantor Fluid Handling
VWR Item#MFLX07003-14
Your Organization's Price:
Extend the life of your gear pump head with these service kits.
  • Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
Service kit includes: gears, suction shoe, shafts, scales, and springs.
More Product Information
Other Products In this Family
See all Items in this Product Family
Learn more
About VWR
Plus About VWR