Micropump L22058 Gear Pump Service Kit for 07001-80; 1/Ea
Fournisseur: Avantor Fluid Handling
Extend the life of your gear pump head with these service kits.
- Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
Service kit includes: gears, suction shoe, shafts, scales, and springs.