Imprimer
BarbLock Assembly Hand Tool for 3/8" tubing; 1/Ea
Fournisseur: Avantor Fluid Handling
VWR Item#MFLX30604-12
Your Organization's Price:
Easily install your Barblock Fittings.
  • Optimize performance of your Barblock fittings
Select a hand tool to assemble your BarbLock Retainers based on the tubing size. Universal removal tool is also available.
<b>Information de livraison: </b>10” plier for the BarbLock hand tools.
More Product Information
Other Products In this Family
See all Items in this Product Family
Learn more
About VWR
Plus About VWR