Imprimer
Viton Transfer Pump Tubing, Viton®, 1/16" ID x 1/8" OD; 25 ft
Fournisseur: Avantor Fluid Handling
VWR Item#MFLX06434-01
Your Organization's Price:
keep light sensitive samples safe.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Acid and solvent transfer; high-temperature work.<br />Characteristics: Opaque black.<br />Certification: Tubing is manufactured according to GMP. <br />Temperature range: –25 to 399 °F (–32 to 204 °C)<br />Sterilize: Not recommended.
More Product Information
Other Products In this Family
See all Items in this Product Family
Learn more
About VWR
Plus About VWR