Imprimer
Maxima™ H minus first strand cDNA synthesis kits
Fournisseur: Thermo Fisher Scientific
K1651 K1652 K1681 K1682
K1651EA 179 EUR
K1651 K1652 K1681 K1682
Maxima™ H minus first strand cDNA synthesis kits
Réactifs pour acides nucléiques RT-PCR and cDNA Reagents
Maxima™ H Minus First Strand cDNA Synthesis Kit is a complete system for highly efficient synthesis of first strand cDNA. The kit uses the Maxima™ H Minus Reverse Transcriptase (RT), which is an advanced enzyme derived by in vitro evolution of M-MuLV RT. The enzyme features the highest thermostability among the derivatives of M-MuLV RT and lacks RNase H activity. The Maxima™ H Minus First Strand cDNA Synthesis Kit allows synthesis of long cDNAs up to 20 kb at elevated temperatures (up to 65 °C), superseding other systems' abilities to produce full-length cDNA. Due to increased synthesis rates the reaction can be completed in 30 minutes.
  • Integrated genomic DNA removal step
  • Increased reaction temperatures: The first strand of cDNA can be synthesised within the 42 to 65 °C temperature range
  • High yields of full-length first strand cDNA with RNA templates up to 20 kb
  • Flexible priming: Oligo(dT)18, random hexamer, or gene-specific primers
Maxima™ H Minus First Strand cDNA Synthesis Kit with dsDNase provides a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. The kit contains a novel double-strand specific DNase (dsDNase) engineered to remove contaminating genomic DNA from RNA preps in 2 minutes without damage to quality or quantity of RNA. Highly specific dsDNase activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved and dsDNase treated RNA can be directly added to reverse transcription.
Information de livraison: Maxima™ H Minus First Strand cDNA Synthesis Kit contains Maxima™ H Minus Enzyme Mix, Oligo(dT)18 and Random hexamer primers, 5X RT Buffer, dNTP Mix, and nuclease-free water. Maxima™ H Minus First Strand cDNA Synthesis Kit with dsDNase, in addition to the above listed components, contains dsDNase and 10X dsDNase Buffer.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Application
Learn more
About VWR
Plus About VWR