Imprimer
Woulff bottles
Fournisseur: witeg Labortechnik
5.802.001
WITG5.802.001EA 183 EUR
WITG5.802.001
Woulff bottles
Flacons Flacons de Woulff
Made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
Two side necks with GL 18 thread, center neck with GL 25 thread. With bottom tubulature with screw cap and olive for tubing connection.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR