Imprimer
Glass density bottles acc. Gay-Lussac
Fournisseur: witeg Labortechnik
3.902.002
WITG3.902.002EA 29.6 EUR
WITG3.902.002
Glass density bottles acc. Gay-Lussac
Flacons de gravité spécifique
Made of borosilicate glass.
Pycnometer with with ground-in capillary stopper, precisely adjusted to the second place of decimal, acc. to ISO 3507.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR