Imprimer
Sulfonation flasks, with four necks
Fournisseur: witeg Labortechnik
0.850.050 0.850.035 0.850.031 0.850.032 0.850.074 0.850.101 0.850.600
WITG0.850.050EA 168 EUR
WITG0.850.050 WITG0.850.035 WITG0.850.031 WITG0.850.032 WITG0.850.074 WITG0.850.101 WITG0.850.600
Sulfonation flasks, with four necks
Ballons Flacons pour sulfonation
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two parallel and two angled side necks.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR