Imprimer
Power selector
Fournisseur: WINKLER
WRT00011-XX010
WINKWRT00011-XX010EA 224 EUR
WINKWRT00011-XX010
Power selector
Contrôleurs Contrôleurs de température
Plug-in power selector made from polystyrene with safety socket with integrated fuse.
  • Rotary switch mechanism with bimetal element
  • Infinitely variable power up to 100%
  • Signal light indicates control status
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR