Imprimer
Septa for Precision NMR tubes, SP Wilmad-LabGlass
Fournisseur: SP Industries
WG-3891-100
WILMWG-3891-100EA 124 EUR
WILMWG-3891-100
Septa for Precision NMR tubes, SP Wilmad-LabGlass
Septa
Natural rubber septa suitable for 5 mm OD thin wall Precision NMR tubes.
  • Designed to maintain excellent glass to rubber contact
  • Suitable for air-sensitive samples, catalytic studies and titration experiments
Colour: white
These septa are manufactured under 'white room' conditions with certified raw materials for consistency.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR