Imprimer
WHEATON® Plastic Media Bottle, PC
Fournisseur: DWK Life Sciences
W017795
WHEAW017795EA 0 EUR
WHEAW017795
WHEATON® Plastic Media Bottle, PC
Flacons Flacons en plastique
These WHEATON® square/octagonal plastic media bottles are manufactured from polycarbonate with blue tamper-evident linerless polypropylene cap attached.
  • Ideal for the storage and transport of media, laboratory reagents, and biological buffers
  • Molded graduations provide a guide for volume filling
  • Cleanroom packaged with multi-barrier elastomer packaging
  • Square footprint ensures efficient use of storage space
  • Autoclavable
These polycarbonate bottles are supplied non-sterile, but can be safely sterilized by gamma irradiation without property loss or
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR