Imprimer
Sample bottles, narrrow neck, with screw cap, WHEATON®
Fournisseur: DWK Life Sciences
990477
WHEA990477EA 79.4 EUR
WHEA990477
Sample bottles, narrrow neck, with screw cap, WHEATON®
Flacons Flacons en verre
Glass, clear.
  • Excellent for accessing fluid flow in deep manholes and hard to reach sub-surface sampling areas
  • Helps meet EPA requirements for water and waste water sampling
  • 38-430 thread
Each sampler consists of an inert polypropylene head with stainless steel fittings and an aluminum pole assembly with a golden alodine finish. It comes with a 1000 mL narrow-mouth sample bottle with a PTFE- lined cap. To use it, screw a sample bottle into the head and lower to the desired depth. Pull the cable at the top of the handle which will lift a spring-More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR