Imprimer
Biochrom Lightwave 3+ with Bluetooth® and Printer
Fournisseur: Biochrom
80-3007-50
WAPR80-3007-50EA 0 EUR
WAPR80-3007-50
Biochrom Lightwave 3+ with Bluetooth® and Printer
Spectrophotomètres et Photomètres Spectrophotomètres UV-Visible
A common sight among educational and industrial laboratories alike, the Lightwave 3 series combines robust diode array optics with a cell holder up to 40 mm pathlength for exceptional results.
  • Built in Software providing fast scanning capabilities
  • Fixed wavelength measurements
  • Kinetics
  • Concentration calculation; OD 600
  • Ability to store up to 90 methods
  • Multi-wavelength mode
  • Equation builder
  • Samples accepted in 10, 20 or 40 mm pathlength cuvettes
  • Export results via USB port
Instrument package options can include inbuilt printer or Bluetooth.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR