Imprimer

Tris-(hydroxyméthyl) aminométhane (TRIS, Trométamol) 99,8-100,5, AnalaR NORMAPUR® zA

Fournisseur: VWR Collection
Attention

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

VWRS103155NEA 0 EUR
VWRS103155N
Tris-(hydroxyméthyl) aminométhane (TRIS, Trométamol) 99,8-100,5, AnalaR NORMAPUR® zA
Tris-(hydroxyméthyl) aminométhane (TRIS, Trométamol)
Formule: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Point d'ébullition: 357 °C (1013 hPa)
Point de fusion: 169…172 °C
Densité: 1.328 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00004679
Numéro CAS: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Assay (calculated on anhydrous) 99.8 - 100.5 %
Appearance White crystalline powder
Identification Passes test
Heavy metals (as Pb) Max. 5 ppm
Ignition residue (SO4) Max. 100 ppm
Insolubility in water Max. 30 ppm
Water Max. 0.1 %
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 5 ppm
As (Arsenic) Max. 0.2 ppm
Ca (Calcium) Max. 1 ppm
Cd (Cadmium) Max. 0.1 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.2 ppm
K (Potassium) Max. 0.3 ppm
Mg (Magnesium) Max. 0.3 ppm
Na (Sodium) Max. 5 ppm
Pb (Lead) Max. 0.1 ppm
Absorbance (280 nm) (0.5 mol/l) Max. 0.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR