Imprimer
Filter for ultrapure water system, arium® CellPlus
Fournisseur: Sartorius
H2O-CUF
VIVAH2O-CUFEA 314 EUR
VIVAH2O-CUF
Filter for ultrapure water system, arium® CellPlus
Filtres Filtres de traitement des liquides
The arium® CellPlus is a sterile packed, point-of-use ultrafilter for efficient removal of endotoxins, RNase, DNase, microorganisms and particles. Retrograde contamination is prevented by the protective bell design made up of polycarbonate. It offers high safety for critical cell culture applications.
  • Effective removal of RNase and DNase
  • Reliable removal of endotoxins
  • High flow rate performance
  • Certified quality
The high grade material enables excellent total throughputs and optimal flow rates. Designed for arium® comfort and arium® mini ultrapure water systems.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR