Imprimer
Desiccators
Fournisseur: VITLAB
326294
VITL326294EA 610 EUR
VITL326294
Desiccators
Dessiccateurs Dessiccateurs en plastique
Ideal dessicators for use in educational laboratories.
  • Lid made from PC is sealed with a neoprene gasket
  • Lower portion made from PP can be filled with desiccants
  • Materials to be dried can be placed on a perforated disc made of PP
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR