Imprimer
Ergonomic Eyepiece-less Stereo Microscope with Integrated Digital Camera, Stabila Bench Stand and Substage Illumination, Mantis Pixo
Fournisseur: VISION ENGINEERING
SMP212
VISISMP212EA 6337 EUR
VISISMP212
Ergonomic Eyepiece-less Stereo Microscope with Integrated Digital Camera, Stabila Bench Stand and Substage Illumination, Mantis Pixo
Microscopes
Mantis Pixo compact ergonomic stereo microscope with integrated digital camera. Easy to use with interchangeable lenses. Bench stand with integrated substage illumination. Magnification from 3 to 15× (objectives to be added).
  • High quality optical stereo image
  • Ergonomic working position
  • Long working distance
  • Integrated high-definition camera
  • Video and image capture
  • On screen annotation
  • Custom overlays
  • Three lens multi-view turret
  • Dynamic 3D lighting
  • White/UV lighting option
Eyepiece-less design. Mantis eyepiece-less technology projects a large highMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR