Imprimer
Cleaning Trolley, VoleoPro
Fournisseur: FHP VILEDA SCS
143724
VILE143724EA 421 EUR
VILE143724
Cleaning Trolley, VoleoPro
Chariots
Compact cleaning trolley, manufactured from a combination out of strong plastic and strong aluminum with 2 anodized elliptical aluminum pillars in the center of the patented V-Shape design.
  • Quick and easy to assemble
  • With a footprint of 0,5 m²
  • Easy to manoeuvre and to store where space is limited
  • In the front are 2x Ø100 mm steel swivel castors with roller bearing and in the rear 2x big Ø150 mm castors with plain bearing for easy maneuverability and easy to pass small obstacles on the floor
VoleoPro Standard includes:
Foldaway bin bag lid incl.More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR