Imprimer
Tubing, for milk and dairy dispensing, Tygon S3™ B-44-4X
Fournisseur: Saint Gobain Life Sciences
AAA00004 AAA00007 AAA00003 AAA00006 AAA00001 AAA00002 AAA00063 AAA00065 AAA00009 AAA00010 AAA00011 AAA00012 AAA00017 AAA00018 AAA00019 AAA00022 AAA00023 AAA00024 AAA00027 AAA00029 AAA00036 AAA00037 AAA00038 AAA00039 AAA00045 AAA00047 AAA00055 AAA00059 AAA00062 AAA00070 AAA00074
VERNAAA00004EA 58.6 EUR
VERNAAA00004 VERNAAA00007 VERNAAA00003 VERNAAA00006 VERNAAA00001 VERNAAA00002 VERNAAA00063 VERNAAA00065 VERNAAA00009 VERNAAA00010 VERNAAA00011 VERNAAA00012 VERNAAA00017 VERNAAA00018 VERNAAA00019 VERNAAA00022 VERNAAA00023 VERNAAA00024 VERNAAA00027 VERNAAA00029 VERNAAA00036 VERNAAA00037 VERNAAA00038 VERNAAA00039 VERNAAA00045 VERNAAA00047 VERNAAA00055 VERNAAA00059 VERNAAA00062 VERNAAA00070 VERNAAA00074
Tubing, for milk and dairy dispensing, Tygon S3™ B-44-4X
Tuyaux Transfer Tubing
Tygon S3™ B-44-4X is a phthalate-free flexible tubing for food and beverage dispensing.
  • Smooth, non-porous bore will not trap particulates or promote bacterial growth
  • Resistant to harsh alkaline cleaners and sanitizers
  • Excellent alternative to rigid piping systems
Temperature resistance: −36 to +74 °C
Hardness (Shore durometer A): 66
Density: 1,21
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR